Versaflo™ Hood Assembly with Inner Collar and Premium Head Suspension, 3M™
Supplier: 3M
7000002347
76308-722CS 134.26 USD
76308-722
Versaflo™ Hood Assembly with Inner Collar and Premium Head Suspension, 3M™
Workwear
3M™ Versaflo™ S-655 Hood Assembly consists of a S-950 Premium Head Suspension and a S-605 Hood.
  • Provides head, face, and shoulder coverage
  • Respiratory seal is a comfortable, knitted inner collar
  • General purpose, cost-effective fabric
It is approved for use with certain 3M™ Powered Air Purifying and Supplied Air Respirator Systems.