DURAN® Tamper Evident Red/Blue Screw Cap GL 45 with Linerless Plug Seal, Qorpak®
Supplier: Qorpak
289201
76308-364CS 62.49 USD
76308-364
DURAN® Tamper Evident Red/Blue Screw Cap GL 45 with Linerless Plug Seal, Qorpak®
Closures Bottle Closures
Seal GL 45 Media Bottles with secure cap.
  • Tamper evident
  • Autoclavable
  • Linerless Plug Seal
Polypropylene material allows maximum temperature of 140 °C with minimum temperature of –40 °C.