Semi-Automated Titrator, METTLER TOLEDO®
Supplier: Mettler Toledo
30470855
76307-656EA 3715 USD
76307-656
Semi-Automated Titrator, METTLER TOLEDO®
Titrators
The EasyPlus Easy EPM performs precise semi-automated titrations with the automated burette, allowing the operator to focus on the color change of samples.
  • Precise semi-automated titrations
  • Error free results
  • Simple operation
  • iTitrate user interface
Perform titrations using the automated burette, focusing on the color change of samples. An immersed dispensing tip avoids drop by drop variations. A variety of calculations are already pre-programmed in the instrument. Simply select your measurement unit, and avoid error-prone manual calculations. Perform your analysis using either the touch screen or the new responsive USB wheel controller. Enjoy its precise control and comfortable operation.