Dosing Device, METTLER TOLEDO®
Supplier: Mettler Toledo
30470856
76307-628EA 3130 USD
76307-628
Dosing Device, METTLER TOLEDO®
Titrators
The EasyPlus sample preparation device is designed to perform accurate, error-free, and fully automated, liquid handling tasks down to 10 µl.
  • Easy dispensing applications
  • Clear controls
  • Simple operation
  • iTitrate user interface
The instrument automatically prepares your standard solutions. Simply input the weight of your sample and define a target concentration. Designed for simplicity, the Easy Dose will start your routine task when you tap on the home screen. Perform your analysis using either the touch screen or the new responsive USB wheel controller. Enjoy its precise control and comfortable operation.