SureSTART™ Screw Caps for Glass Screw Top Vials, 9 mm, Level 1 Everyday Analysis, Thermo Scientific
Supplier: Thermo Scientific
CHSC9-30 CHSC9-10F CHSC9-10T CHSC9-20F CHSC9-40F CHSC9-40T CHSC9-20T CHSC9-30F CHSC9-30T CHSC9-35UBF CHSC9-35UBSF CHSC9-35UBST CHSC9-35UBT
76307-612PK 20.95 USD
76307-612 TSCHSC9-10F TSCHSC9-10T TSCHSC9-20F TSCHSC9-40F TSCHSC9-40T TSCHSC9-20T TSCHSC9-30F TSCHSC9-30T TSCHSC9-35UBF TSCHSC9-35UBSF TSCHSC9-35UBST TSCHSC9-35UBT
SureSTART™ Screw Caps for Glass Screw Top Vials, 9 mm, Level 1 Everyday Analysis, Thermo Scientific
Closures Chromatography Vial Closures
Use Thermo Scientific™ SureSTART™ 9 mm screw caps with screw vials that have a 9 mm opening, including our SureSTART 2 ml Glass Screw Vials and 2 ml polypropylene screw Microvials for <2 ml samples (Level 1). Select level 1 caps for chromatography approved, cost optimized products that are suitable for everyday analyses and compatible with all instrument types.
Choose from polypropylene caps with silicone/PTFE or rubber/TEF septa designed to reduce autosampler needle issues. Pre-slit and bonded septa options are available. Our AVCS™ caps incorporate advanced vial closure system technology to ensure optimal seal compression when closing a vial. Use AVCS screw caps with our SureStop™ vials for effective seals, consistent data, and increased sample throughput.