Round Cover Slips, German Glass, Bellco Glass
Supplier: Bellco Glass
1943-10012A
76305-514CS 98.15 USD
76305-514
Round Cover Slips, German Glass, Bellco Glass
Cover Glasses
These round cover slips are specially developed for fluorescence and confocal microscopy.
  • Provides quality reliability and performance
Ideal for neural tissue cell cultures and hard-to-attach.
Packaging: Packed 1 oz. (28 g) per case.