Ingenio® EZporator® Electroporation System, Mirus Bio
Supplier: Mirus Bio
MIR51000
76304-532EA 5637.66 USD
76304-532
Ingenio® EZporator® Electroporation System, Mirus Bio
Electroporation Systems Electroporators
This system is designed to be used with the Ingenio® Electroporation Solution to provide a cost-effective, robust and open transfection system for many hard-to-transfect cell types.
  • Performance: deliver any nucleic acid to hard-to-transfect, stem and primary cells
  • Simplicity: use a single, universal electroporation solution across all cell types
  • Flexibility: easily optimize electroporation parameters for each cell type
The Ingenio® EZporator® Electroporation System is designed to be used with the Ingenio® Electroporation Solution which is ideal forMore Product Information