Brady® BradyPrinter™ M611 Mobile Printer with Brady Workstation Mobile or Desktop Software, Brady Worldwide
Supplier: Avantor
76304-346EA 905.33 USD
76304-346
Brady® BradyPrinter™ M611 Mobile Printer with Brady Workstation Mobile or Desktop Software, Brady Worldwide
Printers Label Printers
With the BradyPrinter™ M611 mobile printer and the Brady workstation express labels mobile app, you get the complete print-from-everywhere solution that bridges the gap between the office and the field.
  • Maximum label/tape with 2" width
  • Single color printing capability
  • 3.5" display size
  • Thermal transfer print technology
  • Can be used to process maximum 750 labels per day
Mobile label printer - combine the convenience of your mobile device with dependable printing capabilities. Easily create and edit labels in the field with the Brady workstation express labels mobile app. Ideal for wire and cable identification and lab applications. Print directly from your mobile device via bluetooth or Wi-Fi connectivity. Work all day with the rugged design, long-lasting battery and built-in auto-cutter.
Accessories information: Printer Hard Case - designed to carry the BradyPrinter™ M611 printer and extra supplies anywhere you need it.The durable hard case protects your printer against impacts and dust while allowing for easy mobility. Keeps printer accessories organized and on hand.
Printer Magnet - allows printer to affix to panel door or metal surface for more hands-free operation or storage. Attaches to back of printer with screws to ensure stability.
Printer Sling Pack - convenient carryall can be used as a full on-the-go print station. Includes compartments sized to fit your phone, materials and cords so you can keep everything you need together. Has an adjustable padded shoulder strap so you can keep working comfortably. Soft cover protects the printer during transport.