TN300B Series Class 100 Nitrile Glove, TechNiGlove
Supplier: Techniglove
TN300B TN301B TN302B TN303B TN304B TN305B
76302-850CS 433.32 USD
76302-850 76302-852 76302-854 76302-856 76302-858 76302-860
TN300B Series Class 100 Nitrile Glove, TechNiGlove
Gloves
TechNiCleaned Class 100 nitrile gloves are puncture-resistant and 100% nitrile, reducing the potential for allergic reactions to natural ruber protiens.
  • Blue finish
  • Batch specific COC available
These products contain no natural rubber or silicone, and meet the most stringent requirements for particles and extractables. The proprietary microtextured formers offer an excellent grip on all surfaces. These gloves are suitable for use in cleanrooms and contamination controlled work environments, pharmaceutical and medical device manufacturing.
Packaging: Packaged 100 gloves per double poly-sealed bag, 10 bags per case.