Lovibond® Dipslide Incubator, DI 10
Supplier: Lovibond
56B000701
76302-796EA 860.34 USD
76302-796
Lovibond® Dipslide Incubator, DI 10
Incubators Standard Incubators
The Lovibond® DI 10 Incubator is designed for the reliable incubation of bacteriological slides, on-site, in a laboratory or even while mobile in a car or van.
  • Robust design
  • Holds up to 12 dipslides
  • Excellent temperature stability
  • In-car operation
The incubator, when used in conjunction with dipslides, enables effective microbiological monitoring of cooling water in accordance with many European guidelines.
Dipslides provide a crucial part in a testing program, but must be used correctly and regularly as part of a planned regime, week on week to beMore Product Information