Excellence Benchtop Refractometers, Mettler-Toledo
Supplier: Mettler Toledo
30474903 30474904
76300-940EA 14480 USD
76300-940 76300-942
Excellence Benchtop Refractometers, Mettler-Toledo
Refractometers Benchtop Refractometers
Developed for a wide range of applications, the intuitive Excellence Refractometers are the next generation of refractive index measurement.
  • Color touchscreen with 7" display
  • Compatible with nine languages
  • Accuracy up to five digits
  • One Click user interface
  • Simultaneous determination of density, refractive index, pH / conductivity and color
  • Internal storage of up to 200 results
Brix, refractive index and more: METTLER TOLEDO refractometers master measurements of a wide variety of challenging samples with high repeatability and an accuracy of up to five decimal places.
Ordering information: Can be further enhanced with the following Optional modules and accessories to fit your process needs: powerful autosamplers, LabX PC software, ErgoSens infrared motion sensor. Contact your Avantor representative for more information.