Horizon Horizontal Centrifuge, HORIZON 24, Drucker Diagnostics
Supplier: Drucker
00-284-009-000
76300-136EA 4940.01 USD
76300-136
Horizon Horizontal Centrifuge, HORIZON 24, Drucker Diagnostics
Centrifuges Benchtop Centrifuges
The HORIZON 24 routine centrifuge is designed for versatility, large capacity, simplicity, and ease of use. Spin tubes with the simple set-and-lock controls for maximum repeatability. Select from one of three preset cycles with the press of a button, or program your labs custom settings. Lid lights flash throughout the cycle and allow you to easily see the status of the centrifuge.
  • Provides maximum safety and durability
  • Horizontal centrifugation
  • Set and lock to a single setting
  • LED lid lighting indicates status
The convection based cooling system protects the samples even under the heaviest workloads. Lid lights flash at the end of each cycle and continue flashing until the lid is opened. Centrifuge will not run with the lid open and the lid cannot be opened while samples are spinning.