VWR® T Balances
Supplier: VWR International
76300-056 76300-070 76300-072 76300-074 76300-076 76300-078 76300-080 76300-082 76446-794 76446-796 76446-798 76446-800 76446-802 76446-804 76446-808
76300-056EA 6772.27 USD
76300-056 76300-070 76300-072 76300-074 76300-076 76300-078 76300-080 76300-082 76446-794 76446-796 76446-798 76446-800 76446-802 76446-804 76446-808
VWR® T Balances
Balances and Scales
The VWR® T-Series balances feature state-of-the-art technology to provide the highest accuracy measurements.
  • Large, 5" color touch screen
  • Touchless operation via IR proximity sensors
  • Internal and external calibration
  • Seven databases
  • 2× RS232, USB (A and B), ethernet and Wi-Fi communication interfaces
  • Tool-free assembly and disassembly of the three-door draft shield
  • 5-year warranty
These balances provide accuracy and precision for any laboratory environment. Featuring internal calibration, IR proximity sensors for hands free operation, a multitude of communication options, and built in data protections, and provides advanced features for any application. The large, color touch screen allows for easy viewing of results and navigation through the balances menus. Data safety is provided via the internal memory allows for recording up to 500000 weighings. The analytical models feature a spacious 3-door draft shield that allows for tool-free assembly and disassembly of the weighing chamber. With its 5-year warranty, this balance will provide reliable results for years to come.
Certifications: C-UL (US) and CE.
Ordering information: Accessories not included (must be ordered separately).
Delivery information: The 0.01 and 0.1 mg readability models feature a draft shield (shield can't be removed). The 0.001 g readability feature a removable, glass shield. All units supplied with switching AC adapter, weighing pan, weighing pan supports, user manual, and quick start guide. Includes five-year warranty.