Monarch® RNA Cleanup Wash Buffer, New England Biolabs
Supplier: New England Biolabs (NEB)
T2042L
76299-640EA 45 USD
76299-640
Monarch® RNA Cleanup Wash Buffer, New England Biolabs
Buffers Electrophoresis Buffers Washing Buffers
This is a component of the Monarch® RNA cleanup kits, is used to wash RNA after binding to ensure the removal of contaminants such as enzyme, NTPs, and short RNA fragments. It is supplied as a 5X concentrate and requires dilution with ethanol (≥95%) before use.
Monarch® kit components are available separately.
Delivery information: The product is supplied in 2 bottles, each with 20 ml of concentrated buffer.