Monarch® RNA Cleanup Binding Buffer, New England Biolabs
Supplier: New England Biolabs (NEB)
T2041L
76299-638EA
126
USD
76299-638
Monarch® RNA Cleanup Binding Buffer, New England Biolabs
Buffers
Biological Buffers
This buffer is a component of the Monarch® RNA cleanup kits, is a guanidine-based buffer designed to dilute the sample and prepare the RNA sample to bind to the silica columns.
Monarch® kit components are available separately.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)