Rubber Stoppers, 1 Hole, United Scientific Supplies
Supplier: United Scientific Supplies
RST6A-H RST000-H-VWR RST00-H-VWR RST0-H-VWR RST1-H-VWR RST2-H-VWR RST3-H-VWR RST4-H-VWR RST5-H-VWR RST5A-H-VWR RST6-H-VWR RST6A-H-VWR RST7-H-VWR RST7A-H-VWR RST8-H-VWR RST8A-H-VWR RST9-H-VWR RST9A-H-VWR RST10-H-VWR RST10A-H-VWR RST11-H-VWR RST11A-H-VWR RST12-H-VWR RST13-H-VWR RST14-H-VWR RST15-H-VWR RST16-H-VWR
470005-776PK 20 USD
470005-776 76293-666 76293-668 76293-670 76293-672 76293-674 76293-676 76293-678 76293-680 76293-682 76293-684 76293-686 76293-688 76293-690 76293-692 76293-694 76293-696 76293-698 76293-700 76293-702 76293-704 76293-706 76293-708 76293-710 76293-712 76293-714 76293-716
Rubber Stoppers, 1 Hole, United Scientific Supplies
Stoppers Rubber Stoppers
Solid, black stoppers contain a high percentage of pure rubber and weigh less than neoprene.
  • Resilient and pliable for a long time
  • Low in free sulfur
  • Can withstand adverse laboratory conditions without hardening
These black stoppers are made of solid natural rubber that stays pliable over a long period of time. Ideal for use with non-aggressive or concentrated solutions. Low sulfur content. Stoppers will withstand temperatures from –25 to 70 °C (–13 to 158 °F). Each stopper has its size marked in raised figures.
Packaging: Sold as one-pound bags.