Vacuum Adapters, 24/40 Joint, United Scientific Supplies
Supplier: United Scientific Supplies
ADVS01-24-VWR ADV160-24-VWR
76293-228EA 42.91 USD
76293-228 76293-470
Vacuum Adapters, 24/40 Joint, United Scientific Supplies
Adapters Adapters, Conical Ground Joints
Straight and curved vacuum adapters.
  • Straight adapter has a top outer joint and bottom inner joint
  • Bent adapter has a 105° angle between the top outer joint and bottom inner joint
Vacuum adapters have 24/40 joints.
Delivery information: Include a 3/8" hose connector.