Clamp with Hook, United Scientific Supplies
Supplier: United Scientific Supplies
CLHK01-VWR
76293-410EA 11.11 USD
76293-410
Clamp with Hook, United Scientific Supplies
Clamps General Purpose Clamps
Clamp with Hook can be rotated and locked in any position.
  • 10 cm long support rod
Ideal for supporting suspended thermometers, drying tubes and more. The clamp attaches to rods up to 13 mm (1/2") O.D.