Burette Clamp with Boss Head, Uncoated Jaws, United Scientific Supplies
Supplier: United Scientific Supplies
COBR3-U-VWR
76293-408EA 8.11 USD
76293-408
Burette Clamp with Boss Head, Uncoated Jaws, United Scientific Supplies
Clamps General Purpose Clamps
Excellent for holding small objects.
  • Connects to rods up to 13 mm
  • Jaws can be rotated
The jaws open from 15 mm to 45 mm (0.5" to 1.75"). The clamp attaches to rods up to 13 mm (0.5") O.D. Jaws can be rotated and locked at any angle.