Burette Clamps, Polypropylene, United Scientific Supplies
Supplier: United Scientific Supplies
45101-VWR 45102-VWR
76293-400PK 97.71 USD
76293-400 76293-402
Burette Clamps, Polypropylene, United Scientific Supplies
Clamps General Purpose Clamps
Non-corroding, polypropylene plastic clamps hold burettes firmly in place.
  • Corrosion-proof
  • Spring loaded to hold burettes
These polypropylene burette clamps are completely corrosion-proof. The spring-loaded jack in the middle provides a vice like grip on the burette and prevents it from slipping. Two tapered wedges are included to ensure firm grip on 1/2" or 3/8" diameter rods.