3-Prong Extension Clamp, with Rod and PVC-Coated Grips, United Scientific Supplies
Supplier: United Scientific Supplies
ECPVC1-VWR
76293-364EA 21.31 USD
76293-364
3-Prong Extension Clamp, with Rod and PVC-Coated Grips, United Scientific Supplies
Clamps General Purpose Clamps
The deluxe line of 3-prong extension clamps are constructed of pressure die-cast zinc alloy and are chrome-plated.
  • PVC coated Jaws
  • Opens up to 3"
Clamps open to approximately 3", have a recommended maximum grip size of approximately 2.25", and feature double keyed spring-type adjustment screws. Extension rod is 8.25" long, 5/16" O.D., with an overall length of 12.75".