Lattice Lab System Set #2, United Scientific Supplies
Supplier: United Scientific Supplies
LLS002-VWR
76293-334EA 393.99 USD
76293-334
Lattice Lab System Set #2, United Scientific Supplies
Supports Flask Supports
This Lattice Lab System features an assortment of aluminum rods from 18" to 48" long.
  • Sturdy aluminum construction
  • Includes mounting hardware to tables
Assembly is easy and requires only a screwdriver. Support rods are made of hard aluminum, and foot plates and hook connectors are made of cast alloy.
Delivery information: Set #2 includes: (4) Foot Plates, 2.3125" diameter; (4) Set Screws for Foot Plate; (12) Anchoring Screws for Foot Plate; (26) Hook Connectors with Thumbscrew; (8) Support Rods, 2" L, 0.5" diameter; (2) Support Rods, 18" L, 0.5" diameter; (5) Support Rods, 24' L, 0.5" diameter; (2) Support Rods, 36" L, 0.5" diameter; (4) Support Rods, 48" L, 0.5" diameter