16-Piece Organic Chemistry Glassware Kit, United Scientific Supplies
Supplier: United Scientific Supplies
ORGKIT-16-VWR
76293-310EA 335.53 USD
76293-310
16-Piece Organic Chemistry Glassware Kit, United Scientific Supplies
Labware Sets
This 16-piece deluxe set is designed to cover essential requirements for organic chemistry and is suitable for preparations up to 150 g.
  • All glassware is jointed
  • Supplied in a sturdy, form fitting case
This organic chemistry glassware kit comes with a variety of essential parts for conducting experiments.
Delivery information: Kit includes (Joint Sizes): Still head socket 14/23 (cone 19/26), receiver 19/26 (cone 14/23), air leak (cone 14/23), liebig condenser socket 19/26 (cone 19/26), 100 ml dropping funnel (socket 19/26, Cone 19/26), reduction adaptor socket (19/26, cone 24/29), multiple adaptor socket (19/26, cone 24/29), 50 ml round bottom flask (socket 24/29), 250 ml erlenmeyer flask (socket 24/29), stopper (19/26), thermometer pocket (14/23), receiver adaptor socket (19/26), 100 ml round bottom flask (socket 24/29), 250 ml round bottom flask (socket 24/29), 3 stoppers (24/29).