Microscope Storage Cabinet, Diversified Woodcrafts
Supplier: Diversified Woodcrafts
372-4822K
76292-584EA 5107.5 USD
76292-584
Microscope Storage Cabinet, Diversified Woodcrafts
Furniture Storage Cabinets
Constructed of solid oak, hardwood and oak veneers, using dowel construction, with a chemical resistant, earth-friendly UV finish and rubber base molding
  • Adjustable wire dividers all for storage of various size microscopes
  • Holds upto 25 to 30 microscopes
  • Locking oak framed doors with ³/₁₆" tempered glass
  • Shelves are 1" thick and have a weight capacity of 40 lbs. per sq. ft.
Solid oak framed glazed doors are hinged with three-point locking handles. Microscopes are stored safely and conveniently on two fixed and three adjustable shelves. Dividers can be removed to use shelves for general storage.