Seismic-Rated Anchor for Barricade Racks, Justrite®
Supplier: Justrite
35350
76292-556EA 6.73 USD
76292-556
Seismic-Rated Anchor for Barricade Racks, Justrite®
Furniture Storage Cabinets
This Seismic-Rated Anchor for Barricade Racks provides safety when moving and storing of gas cylinders.
  • Listed for use in seismic zones A, B, C, D, E and F
  • For use with barricade style racks
Comprised of a high-strength threaded stud body, expansion clip, nut and washer, these seismic wedge-type anchors are designed for unbroken concrete.
Bodies are made of plated carbon steel, while the expansion clip consists of a split cylindrical ring with undercutting grooves.
Certifications: 2015 IBC Compliant and anchors are tested to ACI 355.2 and ICC-ES AC193.