Swivel Caster Set for Gas Cylinder Hand Trucks, 3", Justrite®
Supplier: Justrite
35376
76292-454EA 39.64 USD
76292-454
Swivel Caster Set for Gas Cylinder Hand Trucks, 3", Justrite®
Carts
This Swivel Caster Set for Gas Cylinder Hand Trucks provides safety when moving and storing of gas cylinders.
Post swivel replacement caster set for single and double cylinder Justrite® hand trucks.
Two solid 3" polypropylene-alloy wheels with ball bearing steel swivel yokes, on a 0.5" threaded post with flat washer, lock washer and nut.