Pneumatic Wheel for Gas Cylinder Hand Trucks, 16", Justrite®
Supplier: Justrite
35396
76292-430EA 142.4 USD
76292-430
Pneumatic Wheel for Gas Cylinder Hand Trucks, 16", Justrite®
Carts
This Pneumatic Wheel for Gas Cylinder Hand Trucks provides safety when moving and storing of gas cylinders.
  • Replacement wheel for double cylinder Justrite® hand trucks
  • Overall size is 16×6".
Features speed-rated four-ply HD construction and a welded steel hub with dual ball bearings, sized for 0.75" axle (not included).