Digi-Sense® Temp14 Single-Input Thermistor Thermometer, Cole-Parmer
Supplier: Antyila Scientific
91428-05
76292-166EA 667.2 USD
76292-166
Digi-Sense® Temp14 Single-Input Thermistor Thermometer, Cole-Parmer
Thermometers Digital Thermometers
Heavy-duty thermometer offers versatility and accuracy in a compact design.
  • Three way hands-free operation available
  • Large easy-to-read screen with back light
  • Rugged armor with built-in stand—withstand the toughest conditions
  • Easy-to-use automatic field calibration ensures accurate readings
  • Low battery indicator
Thermistor measurement type.
Ordering information: Probes are sold separately. Includes protective rubber armor/boot with stand and three AA batteries.