Multi-Barrier Pouches for EasiCollect+ Card, Qiagen
Supplier: Avantor
76291-816PK 152.25 USD
76291-816
Multi-Barrier Pouches for EasiCollect+ Card, Qiagen
Bags Sample Bags
These pouches are specially designed to protect DNA samples collected on FTA EasiCollect+ Cards during transport and storage. DNA sample collection is more efficient when you have the right size pouch to label and protect your samples, from the point of collection, to storage, to the lab.
  • Protect samples from exposure to gas or liquid contamination through seven layers of lamination
  • Tamper-evident seal for forensic DNA sample security
  • Outer paper surface for labelling or writing
  • Same construction as the large multi-barrier pouches in a smaller size for storage of FTA EasiCollect+ Card or other smaller FTA cards
Small multi-barrier pouches maintain sample integrity and security during archiving and transporting of FTA EasiCollect+ Cards and other smaller FTA cards. Seven-layer lamination protects DNA samples from exposure to gas or liquid contamination to ensure sample integrity. With a tamper-evident seal, this pouch is well-suited to forensic DNA sample protection. Multi-barrier pouches are also available in an appropriate size for larger FTA cards.