Provue Solas XL58 Magnifying Lamp, Aven Tools
Supplier: Aven Tools
26501-XL58
76289-848EA 247.07 USD
76289-848
Provue Solas XL58 Magnifying Lamp, Aven Tools
Magnifiers
Equipped with an interchangeable 8-diopter lens.
  • Offers the flexibility of using multiple lenses
  • Fully enclosed spring-balanced arm
  • 60 ultra-bright SMD LEDs with brightness intensity buttons
The Provue Solas XL58 model offers the flexibility of using the included 8-Diopter lens 3× magnification.