Dichloromethane NF, FCC, ACS, Ph. Eur.

Supplier: Greenfield
Warning

313WORLDDM55M
76289-716EA 2553.64 USD
76289-716
Dichloromethane NF, FCC, ACS, Ph. Eur.
Dichloromethane
Formula: CH₂Cl₂
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR