Trio.Bas™ MONO ATEX Kits, Hardy Diagnostics
Supplier: Hardy Diagnostics
BAS207K BAS208K BAS209K BAS210K
76288-354EA 17134.06 USD
76288-354 76288-356 76288-358 76288-360
Trio.Bas™ MONO ATEX Kits, Hardy Diagnostics
Samplers Air Samplers
Hardy Diagnostics introduces the next generation of microbial air samplers for viable particles.
  • Aspirating head with easy manipulation bayonet closure
  • Manual or automatic operations
  • Ergonomic and balanced design to facilitate handling with ease
  • No plugs or external connections
The Trio.Bas™ instruments utilize the impaction method of air sampling. The air, passing through an aspirating head, impacts onto an appropriate agar plate that collects bacteria or other organic substances possibly present in the sampled area. Components and production process have been certified to ATEX standards for Zone 2 explosion hazard areas.
All air samplers are lightweight, with an antibacterial techno-polymer shockproof body, have a long-lasting battery, and are IP65 protected from dust and water. Per USP <797>, Impaction is the preferred method of active air sampling. Use of settling plates for qualitative air sampling cannot be relied upon and shall not be used solely to determine the quality of air in the controlled environment.
Certifications: ISO 14698, USP 797, USP 116, EU GPL-GMP, ISO17025.
Ordering information: Additional accessory components are available separately.
Delivery information: Trio.Bas™ MONO Kit Contains:
Trio.Bas™ MONO, ATEX
Calibration certificate, *Bluetooth data transfer capability
(*Choose desired data download method; separate purchase is required)
Induction battery charger; unit simply rests on top of the base station to charge
Stainless steel aspirating head, autoclavable
Stainless steel cover head, autoclavable
Robust hard-shell carrying case to protect the sampler in transit
IQ, OQ, PQ, fillable documents