EZ-Accu Shot™ Pseudomonas aeruginosa derived from ATCC® 27853™, Microbiologics
Supplier: MICROBIOLOGICS, INC.
0353A
76288-282PK 468.18 USD
76288-282
EZ-Accu Shot™ Pseudomonas aeruginosa derived from ATCC® 27853™, Microbiologics
Controls and Calibration Verification Materials
Quantitative microorganism preparation that delivers less than 100 CFU per 0.1 mL of hydrated suspension.
  • No dilutions required.
Kit contains: 5 pellets of a single enumerated strain, 5 vials of hydrating fluid (1.2 mL per vial)