UVP iBox® Scientia in vivo imaging systems, Analytik Jena
Supplier: Avantor
89237-432EA 118149.17 USD
89237-432 76288-262 76288-264
UVP iBox® Scientia in vivo imaging systems, Analytik Jena
Photodocumentation Systems
In-vivo imaging system detects fluorescent probes in ultra violet to visible to near infrared (NIR) wavelength ranges for small animal imaging.
  • The high resolution and low light sensitivity of the camera allows for faint target detection
  • System comes with a camera choice of either a BioCam 900 or an OptiChemi 615
  • Five position filter wheel
The UVP iBox Scientia systems are easy to use optical imaging systems for noninvasive detection of fluorescent markers in small animals. They are equipped with high resolution, deeply cooled CCD cameras and highly sensitive optics. The high resolution and low light sensitivity of the camera allows for faint target detection. The UVP iBox Scientia comes with epi white, 365 nm UV and 480 nm blue light illumination built into the darkroom. The darkroom also comes with a five position filter wheel with included GFP and RFP emission, excitation filters and a motorized platform with a 10-inch range.
The BioCam 900 has a 1 MP resolution of 1024×1024 with a choice of either a 50 mm f/1.2 or a 25 mm f/0.95 motorized lens. While the OptiChemi 615 has a 3.2 MP (extendable to 9.6 MP) resolution of 2184×1472. It has a choice of either a 50 mm f/1.2 or a 25 mm f/0.95 motorized lens.
The automated eLITE Xe Multispectral Source with external power supply facilitates high intensity illumination with included GFP/RFP excitation filters. The eLITE utilizes a 150 W xenon lamp to provide 400 to 800 nm epi lighting via a fiber optic cable. The eLITE is software controlled and comes with eight excitation filter positions.
Using VisionWorks® Software prioritizes data integrity by gathering unmodified raw image data for research and publications. VisionWorks® Software handles all workflow needs with extensive tools such as: macros, templates, annotations, highlights, text, quantitative analysis, tumor volumes and measurements. VisionWorks® is capable of creating reports and exporting data in order to fully document research. The software supports 21 CFR Part 11 and is compatible with Win 7, 8, and 10.
Certifications: CE certified, CSA listed.
Ordering information: UVP iBox Scientia does not include an Anesthesia kit or a computer.
Delivery information: UVP iBox Scientia includes an eLITE with GFP/RFP filter sets and a warming plate.