Trio.Bas™ CaliTest for Cable Air Sampling Instruments, Hardy Diagnostics
Supplier: Hardy Diagnostics
BAS353
76288-232EA 5857.17 USD
76288-232
Trio.Bas™ CaliTest for Cable Air Sampling Instruments, Hardy Diagnostics
Samplers Air Samplers
A manual verification instrument to check the precision of air flow rates.
  • For control only, not designed to take the place of annual calibrations as required by USP
The CaliTest system is for use only with instruments that have direct data transfer cables.