Vaplock™ Wako 38 mm Waste Caps and Waste Kits, Cole-Parmer
Supplier: Antyila Scientific
12018-68 12018-69 12018-70 12018-71 12018-72 12018-73
76287-756KT 357.56 USD
76287-756 76287-758 76287-760 76287-762 76287-764 76287-766
Vaplock™ Wako 38 mm Waste Caps and Waste Kits, Cole-Parmer
Closures Safety Caps and Safety Waste Caps
The Vaplock™closed system design reduces chemical evaporation and exposure to hazardous vapors.
  • Minimize exposure to harmful chemical vapors
  • Collect solvent waste from multiple effluent streams
  • Highly adaptable and conformable to a broad range of containment types
Featuring a vapor tight connection, waste manifolds are designed to fit standard GL38 threaded lab bottles. Chemical-resistant materials of construction include polypropylene or PTFE manifold and insert, with polypropylene spin collar. These versatile manifolds can be configured for barbed connections, compression fittings, 1/4-28 fittings, and ferrule sets. All manifolds are designed to accommodate the EF-100 Filter and indicator.
Delivery information: Kits include (1) 10-port waste manifold, (3)1/4" NPT polypropylene plugs, (6) 1/4-28 PTFE plugs, (1)1/2" NPT polypropylene plug, (1) organic vapor filter, (2) 2.2 mm O.D. hard wall tubing connections, (1) 2.5 mm O.D. hard wall tubing connection, (3) 1/8" O.D. hard wall tubing connections, (2) 1/16" O.D. hard wall tubing connections, (3) barbed tubing connection 1/4" I.D. tubing×1/4" NPT polypropylene, (1) barbed tubing connection 1/8" I.D. tubing×1/4" NPT polypropylene, (1) barbed tubing connection 3/16" I.D. tubing×1/4" NPT polypropylene, (1) barbed tubing connection 3/8" I.D. tubing×1/4" NPT polypropylene, (1) barb to barb Y connector with 1/4" (6.25 mm) barbs, polypropylene, (1) hard wall tube extension with union connections and 5' of 1/8" O.D. PTFE tubing, (1) filter adapter 1/4" NPT×1/2" NPT polypropylene, and (1) breakthrough color indicator for Vaplock™ closed system organic vapor filter. Container Not Included.