Vaplock™ GL 25 Supply Caps, Cole-Parmer
Supplier: Antyila Scientific
12018-54 12018-55
76287-682EA 53.43 USD
76287-682 76287-684
Vaplock™ GL 25 Supply Caps, Cole-Parmer
Closures Safety Caps and Safety Waste Caps
The Vaplock™ closed system design reduces chemical evaporation and exposure to hazardous vapors with Vaplock™ closed system solvent reservoir caps.
  • Anti-twist design prevents twisting of tubing when the cap is attached or removed as the inner body rotates independently of the outer collar
  • Chemical compatibility with laboratory solvents
Durable port threads are designed to accept industry-standard 1/4-28 threaded fittings for use with semi-rigid tubing equal to or less than 1/8" (3.2 mm) outside diameter.
Chemically compatible caps are constructed of inert PTFE. The body of the cap provides ideal chemical compatibility with laboratory solvents. Under normal use, the outer polypropylene collar is non-wetted. Caps may be autoclaved to 140 °C. Combine with secure fluid connections and Vaplock air inlet or relief valves to maintain pressure equilibrium within the bottle.