First Aid Only Butterfly Bandages, Acme United
Supplier: First Aid Only
G135
76287-288EA 12.42 USD
76287-288
First Aid Only Butterfly Bandages, Acme United
First Aid Supplies Wound Dressings
Butterfly wound closures are latex-free and eliminate the need for stitches.
  • Close and secure small wounds
Butterfly wound closures are designed to close and secure small wounds and incisions while holding the skin firmly. They will not stick to the wound itself. These bandages remove the need for stitches in relatively minor cuts and they are sterile unless package is opened or damaged. Promotes fast healing as well and ensures a secure closure.
Packaging: Packed 100 per box.