Wide Bore Transfer Pipettes, Globe Scientific
Supplier: Globe Scientific
135040-S01
76285-362CS 96.01 USD
76285-362
Wide Bore Transfer Pipettes, Globe Scientific
Pipettes Pasteur/Transfer Pipettes
These wide bore pipettes feature a wide tip opening designed specifically for the transfer of viscous liquids and small particulates.
  • Non-toxic and chemically inert to biological fluids and most acids
  • Easy to grip with gloves
  • Temperature range: –50 to 90 °C
  • Can be gas (EtO) sterilized
Wide bore transfer pipettes are useful in the biofuel, petroleum, water treatment, food/beverage, and pathology industries.