NEBuffer Set (EcoRI, DpnII), New England Biolabs
Supplier: New England Biolabs (NEB)
B7006S
76285-182EA 29 USD
76285-182
NEBuffer Set (EcoRI, DpnII), New England Biolabs
Buffers Cloning Buffers
New England Biolabs provides 10X NEBuffer with each restriction endonuclease to assure optimal (100%) activity.
Set contains two 1.25 ml vials of each unique NEBuffer for the enzymes EcoRI and DpnII. Most of our enzymes are supplied with one of four standard NEBuffers. Occasionally, an enzyme has specific buffer requirements not met by one of the four standard NEBuffers, in which case the enzyme is supplied with its own unique NEBuffer. 1X NEBuffer EcoRI 100 mM Tris-HCl 50 mM NaCl 10 mM MgCl₂ 0.025% Triton X-100 (pH 7.5 @ 25C) 1X NEBuffer DpnII 50 mM Bis-Tris-HCl 100 mM NaCl 10 mM MgCl2 1 mM DTT (pH 6 @ 25C).