Anti-His Tag Mouse Monoclonal Antibody [clone: 3D5]

Supplier: Enzo Life Sciences
ABS537-0200
76285-066EA 639.83 USD
76285-066
Anti-His Tag Mouse Monoclonal Antibody [clone: 3D5]
Antibodies
Recombinant monclonal antibody (Mouse IgG2cκ).

Recommended Applications: ELISA, WB

Type: Primary
Antigen: His Tag
Clonality: Monoclonal
Clone: 3D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2cκ
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR