NEBNext Multiplex Oligos for Illumina (Dual Index Primers Set 2), New England Biolabs
Supplier: New England Biolabs (NEB)
E7780S
76284-672EA 511 USD
76284-672
NEBNext Multiplex Oligos for Illumina (Dual Index Primers Set 2), New England Biolabs
Nucleic Acid Reagents Primers and Probes
NEBNext Multiplex Oligos provide adaptors and primers to enable high yield multiplex Illumina library production. The unique hairpin loop structure of the NEBNext Adaptor minimizes adaptor-dimer formation, and NEBNext index PCR primers enable index incorporation during library amplification.
  • High efficiency adaptor ligation
  • Minimized adaptor-dimer formationIncludes index primers for library multiplexing
  • Increased sample identification specificityIncludes USER Enzyme
Delivery information: This kit includes 8 i5 index primers and 12 i7 index primers for dual indexing.