Anti-PSA Mouse Monoclonal Antibody [clone: 735]

Supplier: Enzo Life Sciences
ABS560-0200
76284-572EA 650.73 USD
76284-572
Anti-PSA Mouse Monoclonal Antibody [clone: 735]
Antibodies
Recombinant monclonal antibody (Mouse IgG2aκ).

Recommended Applications: Flow Cytometry, IHC, IP

Type: Primary
Antigen: PSA
Clonality: Monoclonal
Clone: 735
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2aκ
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR