TSPX2 Innovations Microtome Aligner, Universal, Newcomer Supply
Supplier: Newcomer Supply
6257TO
76278-830EA 369.22 USD
76278-830
TSPX2 Innovations Microtome Aligner, Universal, Newcomer Supply
Microtomes and Microtome Assemblies Microtome Blades and Handles
Universal aligner that aligns any microtome with ease.
  • The microtome aligner can work on all microtomes
  • You only need one unit per lab
  • A must for biopsies
The universal microtome aligner is a must for every histology laboratory. This tool can quickly and easily align a microtome or multiple microtomes to a standard alignment.
Now when you have to do recuts on an aligned microtome, it's efficient and you don't shave away precious tissue trying to get a proper section recut. When you attempting a biopsy recut, this tool is worth its weight in gold.