Statfree® Document Holder Protektive Pak
Supplier: Protektive Pak
47121
76277-264EA 178.17 USD
76277-264
Statfree® Document Holder Protektive Pak
Document Holders
Low charging and dissipative Statfree® Document Holders are used in static sensitive testing and production environments to hold work orders and drawings.
  • Low charging and dissipative, provides path-to-ground
  • Single opening
Imprinted with ESD protective symbol.
Packaging: Sold in packs of 25 each.