Disposal Cabinet, PALBAM
Supplier: PALBAM 2008 AC LTD TE
GDC-8040
76277-164EA 6966.07 USD
76277-164
Disposal Cabinet, PALBAM
Bins Waste Bins
This disposal cabinet can be used on its own as a disposal/recycle unit or it can be combined with dispensers to create a dispensing/disposal unit.
  • Electropolished type 304 stainless steel construction
  • Two waste throw openings, two doors and leveling feet
  • Includes two removable waste bins (plastic) for easy emptying
Can be used in cleanroom environments.