Safety Eyewear, Loctite
Supplier: Loctite
717763
76277-070EA 163.15 USD
76277-070
Safety Eyewear, Loctite
Safety Glasses
UV and visible light safety glasses protect the worker from exposure to wavelengths up to 532 nm.
  • Absorbs 99.9% of potentially harmful UVA and UVB radiation
  • Provides protection by completely absorbing visible light up to 532 nm
The operator contrast is improved and the eye finds it easier to see detail. The orange lens color filters out Visible light in the violet and blue areas of the spectrum.
Certifications: Meets ANSI specifications Z87.1 / CSA Z 94.3.