Labswift NV260 0179 Portable Water Activity Instrument, Cole Parmer
Supplier: Argos Technologies
37910-14
76276-250EA 10037.98 USD
76276-250
Labswift NV260 0179 Portable Water Activity Instrument, Cole Parmer
Hygrometers Digital Thermo-Hygrometers
Portable aw measurement that is quick and easy to perform.
  • Innovative infrared sample temperature measurement
  • Data logger function with SD card interface
  • Test and field calibration functions
  • Sensor filters protect against contamination and errors
  • Spring-loaded sealed measurement chamber
Features data logging, alarm, and digital display.
Certifications: CE Approved, UL Listed.
Delivery information: Supplied with calibration standards (11, 58, 84% RH), 40 sample dishes, 5 prefilters, SD card, tension ring, hard carrying case, external universal power supply, rechargeable lithium-ion battery, and calibration document provided by the manufacturer.