Anti-CALR Mouse Monoclonal Antibody [clone: FMC 75]

Supplier: Enzo Life Sciences
ABS668-0200
76271-380EA 770.63 USD
76271-380
Anti-CALR Mouse Monoclonal Antibody [clone: FMC 75]
Antibodies
Recommended Applications: ICC, IHC (PS), IP, WB

Type: Primary
Antigen: CALR
Clonality: Monoclonal
Clone: FMC 75
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1κ
Reactivity: Human;Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR