Hematoxylin, OPTIK DK type 1

Supplier: Avantik

RS4576-A
76270-186CS 245.37 USD
76270-186
Hematoxylin, OPTIK DK type 1
Hematoxylin

Optik Hematoxylin DK. Uniquely formulated progressive Hematoxylin to produce crisp, well-delineated chromatin while eliminating background staining.


  • Contains no mercury or alcohol
  • Provides a dark, intense stain.
  • 500ml bottle, 4 x 500mL/Cs.

Formula: C₁₆H₁₄O₆
MDL Number: MFCD00078111
CAS Number: 517-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR